Assay Details
Gene Symbol: LOC5569176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845337.1 | XP_021701029.1 | 14 - 15 | 3860 | 86 | |
XM_021845338.1 | XP_021701030.1 | 14 - 15 | 3695 | 86 |
