Assay Details
Gene Symbol: LOC5569176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845337.1 | XP_021701029.1 | 9 - 10 | 2776 | 85 | |
XM_021845338.1 | XP_021701030.1 | 9 - 10 | 2611 | 85 |
