Assay Details
Gene Symbol: LOC5575721 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655776.2 | XP_001655826.2 | 7 - 8 | 2282 | 72 | |
XM_021854152.1 | XP_021709844.1 | 7 - 8 | 2279 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
