Assay Details
Gene Symbol: LOC5575721 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655776.2 | XP_001655826.2 | 3 - 4 | 1391 | 67 | |
XM_021854152.1 | XP_021709844.1 | 3 - 4 | 1391 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
