Assay Details
Gene Symbol: LOC5564567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648865.2 | XP_001648915.2 | 1 - 2 | 115 | 72 | |
GenBank mRNA | AY432478.1 | - | 1 - 2 | 52 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001648865.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
