Assay Details
Gene Symbol: LOC5577908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656632.2 | XP_001656682.1 | 1 - 2 | 119 | ||
GenBank mRNA | AY432488.1 | - | 1 - 2 | 70 | 119 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001656632.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
