Assay Details
Gene Symbol: LOC5577908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656632.2 | XP_001656682.1 | 2 - 3 | 320 | 91 | |
XM_021848191.1 | XP_021703883.1 | 3 - 4 | 397 | 91 | ||
XM_021848192.1 | XP_021703884.1 | 2 - 3 | 497 | 91 | ||
XM_021848193.1 | XP_021703885.1 | 3 - 4 | 533 | 91 | ||
XM_021848194.1 | XP_021703886.1 | 3 - 4 | 406 | 91 | ||
GenBank mRNA | AY432488.1 | - | 2 - 3 | 244 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
