Assay Details
Gene Symbol: LOC5577908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656632.2 | XP_001656682.1 | 3 - 4 | 419 | 71 | |
XM_021848191.1 | XP_021703883.1 | 4 - 5 | 496 | 71 | ||
XM_021848192.1 | XP_021703884.1 | 3 - 4 | 596 | 71 | ||
XM_021848193.1 | XP_021703885.1 | 4 - 5 | 632 | 71 | ||
XM_021848194.1 | XP_021703886.1 | 4 - 5 | 505 | 71 | ||
GenBank mRNA | AY432488.1 | - | 3 - 4 | 343 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
