Assay Details
Gene Symbol: LOC5566949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838683.1 | XP_021694375.1 | 1 - 2 | 237 | 112 | |
XM_021838687.1 | XP_021694379.1 | 1 - 2 | 247 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
