Assay Details
Gene Symbol: LOC5566949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838683.1 | XP_021694375.1 | 3 - 4 | 831 | 120 | |
XM_021838684.1 | XP_021694376.1 | 3 - 4 | 842 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
