Assay Details
Gene Symbol: LOC5566949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838683.1 | XP_021694375.1 | 4 - 5 | 1369 | 55 | |
XM_021838684.1 | XP_021694376.1 | 4 - 5 | 1380 | 55 | ||
XM_021838685.1 | XP_021694377.1 | 3 - 4 | 738 | 55 | ||
XM_021838686.1 | XP_021694378.1 | 3 - 4 | 879 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
