Assay Details
Gene Symbol: LOC23687443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845649.1 | XP_021701341.1 | 8 - 9 | 3831 | 76 | |
GenBank mRNA | AY432533.1 | - | 1 - 2 | 291 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
