Assay Details
Gene Symbol: LOC5568662 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652447.2 | XP_001652497.1 | 2 - 3 | 293 | 102 | |
GenBank mRNA | AY432540.1 | - | 1 - 2 | 549 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
