Assay Details
Gene Symbol: LOC5569547 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652797.2 | XP_001652847.2 | 3 - 4 | 1772 | 58 | |
GenBank mRNA | AY432620.1 | - | 1 - 2 | 23 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
