Assay Details
Gene Symbol: LOC5569547 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652797.2 | XP_001652847.2 | 4 - 5 | 2092 | 64 | |
GenBank mRNA | AY432620.1 | - | 2 - 3 | 343 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
