Assay Details
Gene Symbol: LOC5578974 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841532.1 | XP_021697224.1 | 16 | 3578 | 76 | |
GenBank mRNA | AY432621.1 | - | 1 | 518 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
