Assay Details
Gene Symbol: LOC5576221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655949.2 | XP_001655999.2 | 1 - 2 | 149 | 144 | |
GenBank mRNA | AY432661.1 | - | 1 - 2 | 148 | 144 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001655949.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
