Assay Details
Gene Symbol: LOC5572696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660465.2 | XP_001660515.1 | 2 - 3 | 329 | 77 | |
GenBank mRNA | AY432762.1 | - | 2 - 3 | 310 | 77 | |
DQ440472.1 | - | 2 - 3 | 203 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
