Assay Details
Gene Symbol: LOC5565309 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649607.2 | XP_001649657.2 | 1 - 2 | 2187 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5565309 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649607.2 | XP_001649657.2 | 1 - 2 | 2187 | 69 |
Set Membership: |
![]() ![]() ![]() ![]() |