Assay Details
Gene Symbol: LOC5572845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660553.2 | XP_001660603.1 | 1 - 2 | 325 | 80 | |
XM_021850020.1 | XP_021705712.1 | 3 - 4 | 604 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001660553.2, XM_021850020.1 transcripts.
Set Membership: |
![]() ![]() |
