Assay Details
Gene Symbol: LOC5574609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845802.1 | XP_021701494.1 | 7 - 8 | 3612 | 116 | |
XM_021845803.1 | XP_021701495.1 | 5 - 6 | 2374 | 116 | ||
XM_021845804.1 | XP_021701496.1 | 5 - 6 | 3095 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
