Assay Details
Gene Symbol: LOC5574609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845802.1 | XP_021701494.1 | 6 - 7 | 3456 | 67 | |
XM_021845803.1 | XP_021701495.1 | 4 - 5 | 2218 | 67 | ||
XM_021845804.1 | XP_021701496.1 | 4 - 5 | 2939 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
