Assay Details
Gene Symbol: LOC5574609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845802.1 | XP_021701494.1 | 4 - 5 | 2669 | 70 | |
XM_021845803.1 | XP_021701495.1 | 2 - 3 | 1431 | 70 | ||
XM_021845804.1 | XP_021701496.1 | 2 - 3 | 2152 | 70 |

More Information
Set Membership: |
![]() ![]() |
