Assay Details
Gene Symbol: LOC5574609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845802.1 | XP_021701494.1 | 2 - 3 | 545 | 113 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021845802.1 transcripts.
Set Membership: |
![]() ![]() |
