Assay Details
Gene Symbol: LOC5564337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851565.1 | XP_021707257.1 | 5 - 6 | 768 | 83 | |
XM_021851566.1 | XP_021707258.1 | 4 - 5 | 1124 | 83 |
