Assay Details
Gene Symbol: LOC5564337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851565.1 | XP_021707257.1 | 7 - 8 | 1118 | 81 | |
XM_021851566.1 | XP_021707258.1 | 6 - 7 | 1474 | 81 |
