Assay Details
Gene Symbol: LOC5568816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845633.1 | XP_021701325.1 | 6 - 7 | 1695 | 89 | |
XM_021845634.1 | XP_021701326.1 | 6 - 7 | 1694 | 89 | ||
XM_021845635.1 | XP_021701327.1 | 6 - 7 | 1694 | 89 | ||
XM_021845636.1 | XP_021701328.1 | 6 - 7 | 1694 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
