Assay Details
Gene Symbol: LOC5568816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845635.1 | XP_021701327.1 | 13 - 14 | 4655 | 77 | |
XM_021845636.1 | XP_021701328.1 | 14 - 15 | 4592 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
