Assay Details
Gene Symbol: LOC5565494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649790.2 | XP_001649840.1 | 3 - 4 | 2265 | 81 | |
XM_001649791.2 | XP_001649841.1 | 3 - 4 | 2266 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
