Assay Details
Gene Symbol: LOC5564986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845663.1 | XP_021701355.1 | 8 - 9 | 2580 | 70 | |
XM_021845664.1 | XP_021701356.1 | 8 - 9 | 2561 | 70 | ||
XM_021845665.1 | XP_021701357.1 | 8 - 9 | 2558 | 70 | ||
XM_021845666.1 | XP_021701358.1 | 8 - 9 | 2349 | 70 | ||
XM_021845667.1 | XP_021701359.1 | 9 - 10 | 2852 | 70 | ||
XM_021845668.1 | XP_021701360.1 | 8 - 9 | 2557 | 70 | ||
XM_021845669.1 | XP_021701361.1 | 8 - 9 | 2543 | 70 | ||
XM_021845671.1 | XP_021701363.1 | 8 - 9 | 2502 | 70 | ||
XM_021845672.1 | XP_021701364.1 | 7 - 8 | 1893 | 70 | ||
XM_021845673.1 | XP_021701365.1 | 8 - 9 | 2726 | 70 | ||
XM_021845674.1 | XP_021701366.1 | 10 - 11 | 2392 | 70 | ||
XM_021845675.1 | XP_021701367.1 | 11 - 12 | 2557 | 70 | ||
XM_021845676.1 | XP_021701368.1 | 11 - 12 | 2574 | 70 |

More Information
Set Membership: |
![]() ![]() |
