Assay Details
Gene Symbol: LOC5564986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845663.1 | XP_021701355.1 | 11 - 12 | 3098 | 121 | |
XM_021845664.1 | XP_021701356.1 | 11 - 12 | 3079 | 121 | ||
XM_021845665.1 | XP_021701357.1 | 11 - 12 | 3076 | 121 | ||
XM_021845666.1 | XP_021701358.1 | 11 - 12 | 2867 | 121 | ||
XM_021845667.1 | XP_021701359.1 | 12 - 13 | 3370 | 121 | ||
XM_021845668.1 | XP_021701360.1 | 11 - 12 | 3075 | 121 | ||
XM_021845669.1 | XP_021701361.1 | 11 - 12 | 3061 | 121 | ||
XM_021845671.1 | XP_021701363.1 | 11 - 12 | 3020 | 121 | ||
XM_021845672.1 | XP_021701364.1 | 10 - 11 | 2411 | 121 | ||
XM_021845673.1 | XP_021701365.1 | 11 - 12 | 3244 | 121 | ||
XM_021845674.1 | XP_021701366.1 | 13 - 14 | 2910 | 121 | ||
XM_021845675.1 | XP_021701367.1 | 14 - 15 | 3075 | 121 | ||
XM_021845676.1 | XP_021701368.1 | 14 - 15 | 3092 | 121 |

More Information
Set Membership: |
![]() ![]() |
