Assay Details
Gene Symbol: LOC5564986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845663.1 | XP_021701355.1 | 13 - 14 | 3639 | 79 | |
XM_021845664.1 | XP_021701356.1 | 13 - 14 | 3620 | 79 | ||
XM_021845665.1 | XP_021701357.1 | 13 - 14 | 3617 | 79 | ||
XM_021845666.1 | XP_021701358.1 | 13 - 14 | 3408 | 79 | ||
XM_021845667.1 | XP_021701359.1 | 14 - 15 | 3911 | 79 | ||
XM_021845668.1 | XP_021701360.1 | 13 - 14 | 3616 | 79 | ||
XM_021845669.1 | XP_021701361.1 | 13 - 14 | 3602 | 79 | ||
XM_021845671.1 | XP_021701363.1 | 13 - 14 | 3561 | 79 | ||
XM_021845672.1 | XP_021701364.1 | 12 - 13 | 2952 | 79 | ||
XM_021845673.1 | XP_021701365.1 | 13 - 14 | 3785 | 79 | ||
XM_021845674.1 | XP_021701366.1 | 15 - 16 | 3451 | 79 | ||
XM_021845675.1 | XP_021701367.1 | 16 - 17 | 3616 | 79 | ||
XM_021845676.1 | XP_021701368.1 | 16 - 17 | 3633 | 79 |
