Assay Details
Gene Symbol: LOC5564986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845663.1 | XP_021701355.1 | 15 - 16 | 4055 | 56 | |
XM_021845664.1 | XP_021701356.1 | 15 - 16 | 4036 | 56 | ||
XM_021845665.1 | XP_021701357.1 | 15 - 16 | 4033 | 56 | ||
XM_021845666.1 | XP_021701358.1 | 15 - 16 | 3824 | 56 | ||
XM_021845667.1 | XP_021701359.1 | 16 - 17 | 4327 | 56 | ||
XM_021845668.1 | XP_021701360.1 | 15 - 16 | 4032 | 56 | ||
XM_021845669.1 | XP_021701361.1 | 15 - 16 | 4018 | 56 | ||
XM_021845671.1 | XP_021701363.1 | 15 - 16 | 3977 | 56 | ||
XM_021845672.1 | XP_021701364.1 | 14 - 15 | 3368 | 56 | ||
XM_021845673.1 | XP_021701365.1 | 15 - 16 | 4201 | 56 | ||
XM_021845674.1 | XP_021701366.1 | 17 - 18 | 3867 | 56 | ||
XM_021845675.1 | XP_021701367.1 | 18 - 19 | 4032 | 56 | ||
XM_021845676.1 | XP_021701368.1 | 18 - 19 | 4049 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
