Assay Details
Gene Symbol: LOC5564986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845674.1 | XP_021701366.1 | 1 - 2 | 504 | 80 | |
XM_021845675.1 | XP_021701367.1 | 2 - 3 | 384 | 80 | ||
XM_021845676.1 | XP_021701368.1 | 2 - 3 | 401 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021845674.1, XM_021845675.1, XM_021845676.1 transcripts.
Set Membership: |
![]() ![]() |
