Assay Details
Gene Symbol: LOC5578894 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001664052.2 | XP_001664102.2 | 7 - 8 | 2435 | 66 | |
XM_021844439.1 | XP_021700131.1 | 7 - 8 | 2438 | 66 | ||
XM_021844440.1 | XP_021700132.1 | 7 - 8 | 2390 | 66 | ||
XM_021844441.1 | XP_021700133.1 | 7 - 8 | 2412 | 66 | ||
XM_021844442.1 | XP_021700134.1 | 7 - 8 | 2409 | 66 | ||
XM_021844443.1 | XP_021700135.1 | 7 - 8 | 2383 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
