Assay Details
Gene Symbol: LOC5572863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660573.2 | XP_001660623.2 | 8 - 9 | 2666 | 89 | |
XM_021843703.1 | XP_021699395.1 | 8 - 9 | 2686 | 89 | ||
XM_021843705.1 | XP_021699397.1 | 8 - 9 | 2661 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
