Assay Details
Gene Symbol: LOC5572863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660573.2 | XP_001660623.2 | 6 - 7 | 2218 | 95 | |
XM_021843703.1 | XP_021699395.1 | 6 - 7 | 2238 | 95 | ||
XM_021843705.1 | XP_021699397.1 | 6 - 7 | 2213 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
