Assay Details
Gene Symbol: LOC5572863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660573.2 | XP_001660623.2 | 4 - 5 | 1359 | 94 | |
XM_021843703.1 | XP_021699395.1 | 4 - 5 | 1379 | 94 | ||
XM_021843705.1 | XP_021699397.1 | 4 - 5 | 1354 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
