Assay Details
Gene Symbol: LOC5567813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839879.1 | XP_021695571.1 | 3 - 4 | 961 | 61 | |
XM_021839880.1 | XP_021695572.1 | 3 - 4 | 881 | 61 | ||
XM_021839882.1 | XP_021695574.1 | 2 - 3 | 1186 | 61 | ||
XM_021839883.1 | XP_021695575.1 | 2 - 3 | 1177 | 61 |

More Information
Set Membership: |
![]() ![]() |
