Assay Details
Gene Symbol: LOC5567813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839879.1 | XP_021695571.1 | 4 - 5 | 1098 | 84 | |
XM_021839880.1 | XP_021695572.1 | 4 - 5 | 1018 | 84 | ||
XM_021839882.1 | XP_021695574.1 | 3 - 4 | 1323 | 84 | ||
XM_021839883.1 | XP_021695575.1 | 3 - 4 | 1314 | 84 |
