Assay Details
Gene Symbol: LOC5567813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839879.1 | XP_021695571.1 | 5 - 6 | 1202 | 67 | |
XM_021839880.1 | XP_021695572.1 | 5 - 6 | 1122 | 67 | ||
XM_021839882.1 | XP_021695574.1 | 4 - 5 | 1427 | 67 | ||
XM_021839883.1 | XP_021695575.1 | 4 - 5 | 1418 | 67 |

More Information
Set Membership: |
![]() ![]() |
