Assay Details
Gene Symbol: LOC5567813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839879.1 | XP_021695571.1 | 9 - 10 | 3371 | 86 | |
XM_021839880.1 | XP_021695572.1 | 9 - 10 | 3291 | 86 | ||
XM_021839882.1 | XP_021695574.1 | 8 - 9 | 3596 | 86 | ||
XM_021839883.1 | XP_021695575.1 | 7 - 8 | 2168 | 86 |
