Assay Details
Gene Symbol: LOC5568871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652543.3 | XP_001652593.3 | 16 - 17 | 3117 | 75 | |
XM_021846469.1 | XP_021702161.1 | 16 - 17 | 3118 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
