Assay Details
Gene Symbol: LOC5568871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652543.3 | XP_001652593.3 | 18 - 19 | 3355 | 98 | |
XM_021846463.1 | XP_021702155.1 | 18 - 19 | 3376 | 98 | ||
XM_021846464.1 | XP_021702156.1 | 18 - 19 | 3376 | 98 | ||
XM_021846465.1 | XP_021702157.1 | 17 - 18 | 3199 | 98 | ||
XM_021846466.1 | XP_021702158.1 | 15 - 16 | 3085 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
