Assay Details
Gene Symbol: LOC5568871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652543.3 | XP_001652593.3 | 19 - 20 | 3526 | 93 | |
XM_021846463.1 | XP_021702155.1 | 19 - 20 | 3547 | 93 | ||
XM_021846464.1 | XP_021702156.1 | 19 - 20 | 3547 | 93 | ||
XM_021846465.1 | XP_021702157.1 | 18 - 19 | 3370 | 93 | ||
XM_021846466.1 | XP_021702158.1 | 16 - 17 | 3256 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
