Assay Details
Gene Symbol: LOC5568871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652543.3 | XP_001652593.3 | 21 - 22 | 3845 | 79 | |
XM_021846463.1 | XP_021702155.1 | 21 - 22 | 3866 | 79 | ||
XM_021846464.1 | XP_021702156.1 | 21 - 22 | 3866 | 79 | ||
XM_021846465.1 | XP_021702157.1 | 20 - 21 | 3689 | 79 | ||
XM_021846466.1 | XP_021702158.1 | 18 - 19 | 3575 | 79 | ||
XM_021846467.1 | XP_021702159.1 | 19 - 20 | 3531 | 79 | ||
XM_021846469.1 | XP_021702161.1 | 19 - 20 | 3510 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
