Assay Details
Gene Symbol: LOC5574284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661285.2 | XP_001661335.1 | 2 - 3 | 1173 | 66 | |
XM_021841029.1 | XP_021696721.1 | 2 - 3 | 1427 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
