Assay Details
Gene Symbol: LOC5565563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649859.2 | XP_001649909.1 | 2 - 3 | 742 | 64 | |
XM_021842038.1 | XP_021697730.1 | 2 - 3 | 737 | 64 | ||
XM_021842039.1 | XP_021697731.1 | 2 - 3 | 760 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
