Assay Details
Gene Symbol: LOC5578281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839218.1 | XP_021694910.1 | 3 - 4 | 926 | 62 | |
XM_021839219.1 | XP_021694911.1 | 3 - 4 | 1098 | 62 | ||
XM_021839220.1 | XP_021694912.1 | 3 - 4 | 818 | 62 | ||
XM_021839221.1 | XP_021694913.1 | 3 - 4 | 926 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
