Assay Details
Gene Symbol: LOC5578281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839218.1 | XP_021694910.1 | 4 - 5 | 1025 | 59 | |
XM_021839219.1 | XP_021694911.1 | 4 - 5 | 1197 | 59 | ||
XM_021839220.1 | XP_021694912.1 | 4 - 5 | 917 | 59 | ||
XM_021839221.1 | XP_021694913.1 | 4 - 5 | 1025 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
